Assay Details
Target Gene Details
Entrez Gene ID: | 201305 |
Gene Name: | sphingolipid transporter 3 (putative) |
Gene Aliases: |
- |
Location: |
Chr.17:4433924-4488204 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPNS3 | NM_001320449.1 | NP_001307378.1 | ||
| NM_182538.4 | NP_872344.3 | |||
| XM_011523723.1 | XP_011522025.1 | |||
| XM_011523724.1 | XP_011522026.1 | |||
| XM_011523725.1 | XP_011522027.1 | |||
| XM_011523726.2 | XP_011522028.1 | |||
| AK172832.1 | BAD18797.1 | |||
| BC023646.2 | AAH23646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574268 | Chr.17:4407344 - 4491875 on Build GRCh38 | Gain |
|
| esv3892955 | Chr.17:4310710 - 4476283 on Build GRCh38 | Gain |
|
| nsv574269 | Chr.17:4412211 - 4554285 on Build GRCh38 | Gain |
|
| nsv574267 | Chr.17:4395961 - 4451053 on Build GRCh38 | Loss |
|
| esv3639779 | Chr.17:4412812 - 4481920 on Build GRCh38 | Gain |
|
| esv2762544 | Chr.17:4178680 - 4476552 on Build GRCh38 | Gain |
|
| nsv833345 | Chr.17:4328495 - 4508526 on Build GRCh38 | Loss |
|
| dgv3087n100 | Chr.17:4391668 - 4459140 on Build GRCh38 | Loss |
|
| esv3639773 | Chr.17:4304329 - 4486263 on Build GRCh38 | Gain |
|
| dgv5430n54 | Chr.17:4416311 - 4479434 on Build GRCh38 | Gain |
|
| nsv1058176 | Chr.17:4189086 - 4502476 on Build GRCh38 | Gain |
|
| nsv1065759 | Chr.17:4405261 - 4476552 on Build GRCh38 | Gain |
|
| esv3639781 | Chr.17:4438234 - 4463145 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114948771] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map