Assay Details
Target Gene Details
Entrez Gene ID: | 9772 |
Gene Name: | transmembrane protein 94 |
Gene Aliases: |
KIAA0195 |
Location: |
Chr.17:75441159-75500452 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM94 | NM_001321149.1 | NP_001308078.1 | ||
| NM_014738.5 | NP_055553.3 | |||
| XM_011525514.1 | XP_011523816.1 | |||
| XM_011525515.1 | XP_011523817.1 | |||
| XM_011525516.1 | XP_011523818.1 | |||
| XM_011525517.1 | XP_011523819.1 | |||
| XM_017025440.1 | XP_016880929.1 | |||
| XM_017025449.1 | XP_016880938.1 | |||
| AK304819.1 | ||||
| AK308160.1 | ||||
| BC045540.1 | AAH45540.1 | |||
| D83779.1 | ||||
| DA193442.1 | ||||
| DA317199.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470603 | Chr.17:75468213 - 75741366 on Build GRCh38 | Loss |
|
| esv3641265 | Chr.17:75465751 - 75477877 on Build GRCh38 | Gain |
|
| nsv516133 | Chr.17:75420733 - 75763223 on Build GRCh38 | Loss |
|
| nsv576027 | Chr.17:75249939 - 75750289 on Build GRCh38 | Gain |
|
| nsv833542 | Chr.17:75441043 - 75544993 on Build GRCh38 | Loss |
|

Genomic Map