Assay Details
Target Gene Details
Entrez Gene ID: | 101927557 |
Gene Name: | uncharacterized LOC101927557 |
Gene Aliases: |
- |
Location: |
Chr.17:57522248-57523597 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927557 | NR_110808.1 | |||
| AK131022.1 |
Target Gene Details
Entrez Gene ID: | 124540 |
Gene Name: | musashi RNA binding protein 2 |
Gene Aliases: |
MSI2H |
Location: |
Chr.17:57255851-57684689 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSI2 | NM_001322250.1 | NP_001309179.1 | ||
| NM_001322251.1 | NP_001309180.1 | |||
| NM_138962.3 | NP_620412.1 | |||
| NM_170721.1 | NP_733839.1 | |||
| XM_005257014.3 | XP_005257071.1 | |||
| XM_005257015.3 | XP_005257072.1 | |||
| XM_011524286.1 | XP_011522588.1 | |||
| XM_011524287.2 | XP_011522589.1 | |||
| XM_017024147.1 | XP_016879636.1 | |||
| XM_017024148.1 | XP_016879637.1 | |||
| XM_017024149.1 | XP_016879638.1 | |||
| AK093888.1 | BAC04244.1 | |||
| AK295060.1 | ||||
| AK297295.1 | ||||
| BC001526.2 | AAH01526.1 | |||
| BC017560.2 | AAH17560.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv955e212 | Chr.17:57517630 - 57523908 on Build GRCh38 | Loss |
|
| nsv833496 | Chr.17:57424459 - 57615914 on Build GRCh38 | Gain |
|
| esv33941 | Chr.17:57012706 - 57716106 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map