Assay Details
Target Gene Details
Entrez Gene ID: | 677769 |
Gene Name: | small Cajal body-specific RNA 17 |
Gene Aliases: |
U91 |
Location: |
Chr.18:49814023-49814443 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCARNA17 | NR_003003.2 | 1 | 150 |
Target Gene Details
Entrez Gene ID: | 103091864 |
Gene Name: | small nucleolar RNA host gene 22 |
Gene Aliases: |
SCARNA17, SCARNA17HG |
Location: |
Chr.18:49814023-49851059 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNHG22 | NR_117096.1 | |||
| AK123464.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516056 | Chr.18:49768512 - 49838635 on Build GRCh38 | Gain |
|
| nsv576863 | Chr.18:49569117 - 49938639 on Build GRCh38 | Gain |
|

Genomic Map