Assay Details
Target Gene Details
Entrez Gene ID: | 83473 |
Gene Name: | katanin catalytic subunit A1 like 2 |
Gene Aliases: |
- |
Location: |
Chr.18:46917602-47104795 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KATNAL2 | NM_031303.2 | NP_112593.2 | ||
| XM_005258357.4 | XP_005258414.1 | |||
| XM_005258358.4 | XP_005258415.1 | |||
| XM_006722554.3 | XP_006722617.1 | |||
| XM_011526219.2 | XP_011524521.1 | |||
| XM_011526220.1 | XP_011524522.1 | |||
| XM_011526221.2 | XP_011524523.1 | |||
| XM_011526223.2 | XP_011524525.1 | |||
| XM_017026028.1 | XP_016881517.1 | |||
| XM_017026029.1 | XP_016881518.1 | |||
| XM_017026030.1 | XP_016881519.1 | |||
| XM_017026032.1 | XP_016881521.1 | |||
| AK309134.1 | ||||
| BC034999.2 | AAH34999.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583040 | Chr.18:46955975 - 46966752 on Build GRCh38 | Loss |
|
| nsv509693 | Chr.18:46942006 - 47047880 on Build GRCh38 | Insertion |
|
| dgv3346n100 | Chr.18:46920296 - 46959260 on Build GRCh38 | Loss |
|
| nsv833639 | Chr.18:46846400 - 47026348 on Build GRCh38 | Loss |
|
| nsv9629 | Chr.18:46684730 - 47231140 on Build GRCh38 | Gain+Loss |
|
| nsv1056744 | Chr.18:46929089 - 47055381 on Build GRCh38 | Gain |
|
| esv3574009 | Chr.18:46944650 - 46959260 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map