Assay Details
Target Gene Details
Entrez Gene ID: | 10928 |
Gene Name: | ralA binding protein 1 |
Gene Aliases: |
RIP1, RLIP1, RLIP76 |
Location: |
Chr.18:9475532-9538108 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RALBP1 | NM_006788.3 | NP_006779.1 | ||
| BQ224100.1 | ||||
| BX352266.2 | ||||
| L42542.1 | AAB00103.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833585 | Chr.18:9401632 - 9597293 on Build GRCh38 | Gain+Loss |
|
| nsv1056407 | Chr.18:9144347 - 9553766 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map