Assay Details
Target Gene Details
Entrez Gene ID: | 105371998 |
Gene Name: | uncharacterized LOC105371998 |
Gene Aliases: |
- |
Location: |
Chr.18:12432774-12447798 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105371998 | NR_136514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060808 | Chr.18:12302793 - 12514418 on Build GRCh38 | Gain |
|
| nsv576496 | Chr.18:12247873 - 12490503 on Build GRCh38 | Gain |
|
| nsv1063125 | Chr.18:12046893 - 12918252 on Build GRCh38 | Loss |
|
| esv3380733 | Chr.18:12437853 - 12440151 on Build GRCh38 | Duplication |
|
| esv2758714 | Chr.18:12240682 - 12499526 on Build GRCh38 | Loss |
|

Genomic Map