Assay Details
Target Gene Details
Entrez Gene ID: | 83473 |
Gene Name: | katanin catalytic subunit A1 like 2 |
Gene Aliases: |
- |
Location: |
Chr.18:46917602-47104795 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KATNAL2 | XM_005258357.4 | XP_005258414.1 | ||
| XM_005258358.4 | XP_005258415.1 | |||
| XM_006722554.3 | XP_006722617.1 | |||
| XM_011526219.2 | XP_011524521.1 | |||
| XM_011526220.1 | XP_011524522.1 | |||
| XM_011526221.2 | XP_011524523.1 | |||
| XM_011526223.2 | XP_011524525.1 | |||
| XM_017026028.1 | XP_016881517.1 | |||
| XM_017026029.1 | XP_016881518.1 | |||
| XM_017026030.1 | XP_016881519.1 | |||
| XM_017026032.1 | XP_016881521.1 | |||
| AK309134.1 |
Target Gene Details
Entrez Gene ID: | 9063 |
Gene Name: | protein inhibitor of activated STAT 2 |
Gene Aliases: |
ARIP3, DIP, MIZ, MIZ1, PIASX, PIASX-ALPHA, PIASX-BETA, SIZ2, ZMIZ4 |
Location: |
Chr.18:46803224-46920167 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIAS2 | NM_001324060.1 | NP_001310989.1 | ||
| XM_005258377.3 | XP_005258434.1 | |||
| XM_006722571.3 | XP_006722634.1 | |||
| XM_006722572.3 | XP_006722635.1 | |||
| XM_006722573.2 | XP_006722636.1 | |||
| AF361054.1 | AAK48938.1 | |||
| DB024780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059378 | Chr.18:46851869 - 46929089 on Build GRCh38 | Gain |
|
| nsv522397 | Chr.18:46871681 - 46936125 on Build GRCh38 | Loss |
|
| nsv833639 | Chr.18:46846400 - 47026348 on Build GRCh38 | Loss |
|
| nsv9629 | Chr.18:46684730 - 47231140 on Build GRCh38 | Gain+Loss |
|
| esv2763189 | Chr.18:46835816 - 46935421 on Build GRCh38 | Gain |
|
| esv3583037 | Chr.18:46901568 - 46955974 on Build GRCh38 | Loss |
|
| nsv576813 | Chr.18:46901678 - 46951320 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map