Assay Details
Target Gene Details
Entrez Gene ID: | 23253 |
Gene Name: | ankyrin repeat domain 12 |
Gene Aliases: |
ANCO-2, ANCO1, GAC-1, Nbla00144 |
Location: |
Chr.18:9136753-9285985 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056407 | Chr.18:9144347 - 9553766 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs138720271] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map