Assay Details
Target Gene Details
Entrez Gene ID: | 4645 |
Gene Name: | myosin VB |
Gene Aliases: |
- |
Location: |
Chr.18:49822786-50195081 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 29 - Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYO5B | NM_001080467.2 | NP_001073936.1 | ||
| AB032945.2 | ||||
| AB290160.1 | ||||
| AK025336.1 | BAB15114.1 | |||
| AK093149.1 | ||||
| AK301976.1 | ||||
| AL133643.1 | ||||
| AY274809.1 | AAP42074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576865 | Chr.18:49832262 - 49878828 on Build GRCh38 | Loss |
|
| nsv576863 | Chr.18:49569117 - 49938639 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map