Assay Details
Target Gene Details
Entrez Gene ID: | 2235 |
Gene Name: | ferrochelatase |
Gene Aliases: |
EPP, FCE |
Location: |
Chr.18:57544841-57586737 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FECH | NM_000140.3 | NP_000131.2 | ||
| NM_001012515.2 | NP_001012533.1 | |||
| XM_011525881.1 | XP_011524183.1 | |||
| XM_011525882.2 | XP_011524184.1 | |||
| XM_017025614.1 | XP_016881103.1 | |||
| AK092416.1 | BAC03882.1 | |||
| AK223190.1 | BAD96910.1 | |||
| AK292937.1 | ||||
| AK299643.1 | ||||
| BC039841.1 | AAH39841.2 | |||
| BG772545.1 | ||||
| BT019958.1 | AAV38761.1 | |||
| BT019959.1 | AAV38762.1 | |||
| BX571744.1 | CAE11869.1 | |||
| D00726.1 | BAA00628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833663 | Chr.18:57549030 - 57718589 on Build GRCh38 | Gain |
|
| nsv577032 | Chr.18:57517516 - 57756707 on Build GRCh38 | Gain |
|
| esv2751777 | Chr.18:57515803 - 57805084 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map