Assay Details
Target Gene Details
Entrez Gene ID: | 3613 |
Gene Name: | inositol monophosphatase 2 |
Gene Aliases: |
- |
Location: |
Chr.18:11981428-12030886 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IMPA2 | NM_014214.2 | NP_055029.1 | ||
| XM_011525661.2 | XP_011523963.1 | |||
| AF014398.2 | AAB70915.1 | |||
| AF200432.1 | AAF07824.1 | |||
| BC017176.1 | AAH17176.1 | |||
| BC031560.1 | AAH31560.1 | |||
| BT007061.1 | AAP35710.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758713 | Chr.18:11897024 - 12076103 on Build GRCh38 | Loss |
|
| nsv528699 | Chr.18:11989417 - 12029858 on Build GRCh38 | Loss |
|
| esv3893075 | Chr.18:11989824 - 12147925 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73404704] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map