Assay Details
Target Gene Details
Entrez Gene ID: | 90338 |
Gene Name: | zinc finger protein 160 |
Gene Aliases: |
F11, HKr18, HZF5, KR18 |
Location: |
Chr.19:53064882-53103434 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv580068 | Chr.19:53091553 - 53120518 on Build GRCh38 | Gain |
|
| esv2718787 | Chr.19:52570219 - 53366454 on Build GRCh38 | Deletion |
|
| nsv428370 | Chr.19:52907206 - 53149322 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map