Assay Details
Target Gene Details
Entrez Gene ID: | 5566 |
Gene Name: | protein kinase cAMP-activated catalytic subunit alpha |
Gene Aliases: |
PKACA, PPNAD4 |
Location: |
Chr.19:14091688-14117747 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKACA | NM_001304349.1 | NP_001291278.1 | ||
| NM_002730.3 | NP_002721.1 | |||
| NM_207518.2 | NP_997401.1 | |||
| XM_017026948.1 | XP_016882437.1 | |||
| AK131561.1 | ||||
| AK290147.1 | ||||
| AK292284.1 | ||||
| BC039846.1 | AAH39846.1 | |||
| BC108259.1 | AAI08260.1 | |||
| X07767.1 | CAA30597.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578645 | Chr.19:14086937 - 14116789 on Build GRCh38 | Loss |
|
| nsv833761 | Chr.19:13998659 - 14136778 on Build GRCh38 | Loss |
|
| nsv953980 | Chr.19:14082589 - 14210288 on Build GRCh38 | Deletion |
|
| dgv49n68 | Chr.19:14066502 - 14274131 on Build GRCh38 | Loss |
|
| nsv828452 | Chr.19:14059742 - 14148154 on Build GRCh38 | Loss |
|
| nsv828454 | Chr.19:14061711 - 14096781 on Build GRCh38 | Gain |
|
| nsv578642 | Chr.19:14028192 - 14187630 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114153875] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map