Assay Details
Target Gene Details
Entrez Gene ID: | 57622 |
Gene Name: | leucine rich repeat and fibronectin type III domain containing 1 |
Gene Aliases: |
SALM2 |
Location: |
Chr.19:39306817-39320867 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRFN1 | NM_020862.1 | 2 | 2305 | NP_065913.1 |
| XM_005259102.4 | 5 | 2505 | XP_005259159.1 | |
| XM_017027033.1 | 5 | 2620 | XP_016882522.1 | |
| AB040917.1 | 2 | 2094 | BAA96008.1 | |
| BC025310.1 | 2 | 1618 | AAH25310.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833830 | Chr.19:39218898 - 39347206 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map