Assay Details
Target Gene Details
Entrez Gene ID: | 100505681 |
Gene Name: | NAPA antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:47484282-47501597 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAPA-AS1 | NR_038452.1 | |||
| AK057858.1 | ||||
| BG035979.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758765 | Chr.19:47354499 - 47522248 on Build GRCh38 | Loss |
|
| nsv1056887 | Chr.19:47481963 - 47527369 on Build GRCh38 | Gain |
|
| nsv953591 | Chr.19:47486744 - 47500943 on Build GRCh38 | Deletion |
|
| nsv579888 | Chr.19:47428355 - 47658874 on Build GRCh38 | Gain |
|

Genomic Map