Assay Details
Target Gene Details
Entrez Gene ID: | 83475 |
Gene Name: | deoxyhypusine hydroxylase/monooxygenase |
Gene Aliases: |
HLRC1, hDOHH |
Location: |
Chr.19:3490821-3500940 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOHH | NM_001145165.1 | NP_001138637.1 | ||
| NM_031304.4 | NP_112594.1 | |||
| XM_011528341.2 | 4 | 728 | XP_011526643.1 | |
| BC002817.2 | AAH02817.1 | |||
| BC009863.2 | AAH09863.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953948 | Chr.19:3171703 - 3700002 on Build GRCh38 | Deletion |
|
| nsv470114 | Chr.19:3372516 - 3855979 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map