Assay Details
Target Gene Details
Entrez Gene ID: | 7089 |
Gene Name: | transducin like enhancer of split 2 |
Gene Aliases: |
ESG, ESG2, GRG2 |
Location: |
Chr.19:2997638-3047635 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLE2 | NM_001144761.1 | NP_001138233.1 | ||
| NM_001144762.1 | NP_001138234.1 | |||
| NM_001300846.1 | NP_001287775.1 | |||
| NM_003260.4 | NP_003251.2 | |||
| XM_006722864.2 | XP_006722927.1 | |||
| XM_011528230.1 | XP_011526532.1 | |||
| AK123183.1 | ||||
| AK293407.1 | ||||
| AK293482.1 | ||||
| AK308137.1 | ||||
| AK312285.1 | ||||
| BC017364.2 | AAH17364.1 | |||
| M99436.1 | AAA61193.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833709 | Chr.19:2950244 - 3068609 on Build GRCh38 | Loss |
|
| dgv47n68 | Chr.19:2977776 - 3162121 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map