Assay Details
Target Gene Details
Entrez Gene ID: | 58509 |
Gene Name: | cactin, spliceosome C complex subunit |
Gene Aliases: |
C19orf29, NY-REN-24, fSAPc |
Location: |
Chr.19:3610628-3626815 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACTIN | NM_001080543.1 | 2 | 509 | NP_001074012.1 |
| NM_021231.1 | 2 | 509 | NP_067054.1 | |
| XM_011528160.2 | 2 | 504 | XP_011526462.1 | |
| XM_011528161.2 | 2 | 504 | XP_011526463.1 | |
| AW027691.1 | ||||
| AY917150.1 | 2 | 511 | ||
| CN305928.1 | 2 | 512 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953948 | Chr.19:3171703 - 3700002 on Build GRCh38 | Deletion |
|
| nsv833713 | Chr.19:3564423 - 3627361 on Build GRCh38 | Loss |
|
| nsv470114 | Chr.19:3372516 - 3855979 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map