Assay Details
Target Gene Details
Entrez Gene ID: | 102238594 |
Gene Name: | SPACA6P antisense RNA |
Gene Aliases: |
LINC01129 |
Location: |
Chr.19:51685363-51693456 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPACA6P-AS | NR_108100.1 | 2 | 2373 | |
| AK125996.1 | 2 | 2373 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9742 | Chr.19:51684285 - 51718489 on Build GRCh38 | Gain+Loss |
|
| esv2758767 | Chr.19:51488642 - 52101372 on Build GRCh38 | Gain+Loss |
|

Genomic Map