Assay Details
Target Gene Details
Entrez Gene ID: | 84807 |
Gene Name: | NFKB inhibitor delta |
Gene Aliases: |
IkappaBNS, TA-NFKBH |
Location: |
Chr.19:35887630-35902301 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFKBID | NM_139239.2 | NP_640332.1 | ||
| XM_005259340.3 | XP_005259397.1 | |||
| XM_011527419.2 | XP_011525721.1 | |||
| XM_011527421.2 | XP_011525723.1 | |||
| XM_017027389.1 | XP_016882878.1 | |||
| XM_017027390.1 | XP_016882879.1 | |||
| XM_017027391.1 | XP_016882880.1 | |||
| XM_017027396.1 | XP_016882885.1 | |||
| AF385433.1 | AAM43834.1 | |||
| AF385434.1 | AAM43835.1 | |||
| BX377126.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv51n68 | Chr.19:35841011 - 36016692 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map