Assay Details
Target Gene Details
Entrez Gene ID: | 4606 |
Gene Name: | myosin binding protein C, fast type |
Gene Aliases: |
MYBPC, MYBPCF |
Location: |
Chr.19:50432903-50466326 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYBPC2 | NM_004533.3 | NP_004524.3 | ||
| BC130536.1 | ||||
| BC136389.1 | ||||
| CF552888.1 | ||||
| X73113.1 | CAA51544.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066389 | Chr.19:50319151 - 50453251 on Build GRCh38 | Gain |
|
| esv2661463 | Chr.19:50420747 - 50453759 on Build GRCh38 | Deletion |
|
| nsv509750 | Chr.19:50443078 - 50540357 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111856587] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map