Assay Details
Target Gene Details
Entrez Gene ID: | 170961 |
Gene Name: | ankyrin repeat domain 24 |
Gene Aliases: |
- |
Location: |
Chr.19:4182618-4224814 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD24 | NM_133475.1 | NP_597732.1 | ||
| XM_011527756.2 | XP_011526058.1 | |||
| XM_011527757.2 | XP_011526059.1 | |||
| XM_011527758.2 | XP_011526060.1 | |||
| XM_011527761.1 | XP_011526063.1 | |||
| XM_011527764.2 | XP_011526066.1 | |||
| XM_011527765.2 | XP_011526067.1 | |||
| XM_017026438.1 | XP_016881927.1 | |||
| XM_017026439.1 | XP_016881928.1 | |||
| XM_017026440.1 | XP_016881929.1 | |||
| AB075861.1 | BAB85567.1 | |||
| BI914435.1 | ||||
| CN286920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833716 | Chr.19:4121259 - 4274854 on Build GRCh38 | Loss |
|

Genomic Map