Assay Details
Target Gene Details
Entrez Gene ID: | 57664 |
Gene Name: | pleckstrin homology domain containing A4 |
Gene Aliases: |
PEPP1 |
Location: |
Chr.19:48837092-48868627 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHA4 | NM_001161354.1 | NP_001154826.1 | ||
| NM_020904.2 | NP_065955.2 | |||
| XM_005259107.2 | XP_005259164.1 | |||
| XM_006723301.2 | XP_006723364.1 | |||
| XM_011527157.1 | XP_011525459.1 | |||
| XM_011527158.2 | XP_011525460.1 | |||
| XM_011527159.1 | XP_011525461.1 | |||
| XM_011527160.1 | XP_011525462.1 | |||
| XM_011527162.2 | XP_011525464.1 | |||
| XM_017027037.1 | XP_016882526.1 | |||
| AB208908.1 | BAD92145.1 | |||
| AY007233.1 | AAG01896.1 | |||
| BC024157.2 | AAH24157.1 | |||
| BC033832.1 | AAH33832.1 | |||
| BC064601.1 | AAH64601.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv833855 | Chr.19:48750098 - 48949781 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map