Assay Details
Target Gene Details
Entrez Gene ID: | 56928 |
Gene Name: | signal peptide peptidase like 2B |
Gene Aliases: |
IMP-4, IMP4, PSH4, PSL1 |
Location: |
Chr.19:2328571-2355102 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPPL2B | NM_001077238.1 | NP_001070706.1 | ||
| NM_152988.2 | NP_694533.1 | |||
| XM_011528138.2 | XP_011526440.1 | |||
| XM_017027016.1 | XP_016882505.1 | |||
| XM_017027017.1 | XP_016882506.1 | |||
| XM_017027019.1 | XP_016882508.1 | |||
| AB040965.1 | ||||
| AJ345027.1 | CAC87788.1 | |||
| AJ420897.1 | CAD13134.1 | |||
| AK308973.1 | ||||
| AK309122.1 | ||||
| AY169315.1 | AAO12540.1 | |||
| BC001788.1 | ||||
| BC028391.2 | AAH28391.2 | |||
| BC093046.1 | AAH93046.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160562 | Chr.19:2328434 - 2460264 on Build GRCh38 | Deletion |
|
| esv3643422 | Chr.19:2314254 - 2337163 on Build GRCh38 | Gain |
|
| dgv6218n54 | Chr.19:2221793 - 2336009 on Build GRCh38 | Loss |
|
| nsv953944 | Chr.19:2283702 - 2525802 on Build GRCh38 | Deletion |
|
| nsv833707 | Chr.19:2187092 - 2372223 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80292256] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map