Assay Details
Target Gene Details
Entrez Gene ID: | 23217 |
Gene Name: | zinc finger RNA binding protein 2 |
Gene Aliases: |
KIAA1086 |
Location: |
Chr.19:3804024-3869029 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFR2 | NM_015174.1 | 19 | 2702 | NP_055989.1 |
| XM_011527831.2 | 19 | 2704 | XP_011526133.1 | |
| AB029009.1 | 18 | 2580 | BAA83038.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6227n54 | Chr.19:3804703 - 3842329 on Build GRCh38 | Gain |
|
| nsv833715 | Chr.19:3786758 - 3977144 on Build GRCh38 | Loss |
|
| esv3643475 | Chr.19:3805735 - 3814064 on Build GRCh38 | Loss |
|
| esv3643473 | Chr.19:3789080 - 3847883 on Build GRCh38 | Gain |
|
| nsv509718 | Chr.19:3676947 - 3844683 on Build GRCh38 | Insertion |
|
| nsv470114 | Chr.19:3372516 - 3855979 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map