Assay Details
Target Gene Details
Entrez Gene ID: | 51690 |
Gene Name: | LSM7 homolog, U6 small nuclear RNA and mRNA degradation associated |
Gene Aliases: |
YNL147W |
Location: |
Chr.19:2321521-2328615 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LSM7 | NM_016199.2 | NP_057283.1 | ||
| XM_017026869.1 | XP_016882358.1 | |||
| AF182293.1 | AAD56231.1 | |||
| BC018621.1 | AAH18621.1 | |||
| BI553911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3643422 | Chr.19:2314254 - 2337163 on Build GRCh38 | Gain |
|
| dgv6218n54 | Chr.19:2221793 - 2336009 on Build GRCh38 | Loss |
|
| nsv953944 | Chr.19:2283702 - 2525802 on Build GRCh38 | Deletion |
|
| nsv833707 | Chr.19:2187092 - 2372223 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map