Assay Details
Target Gene Details
Entrez Gene ID: | 8612 |
Gene Name: | phospholipid phosphatase 2 |
Gene Aliases: |
LPP2, PAP-2c, PAP2-g, PPAP2C |
Location: |
Chr.19:281040-291435 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLPP2 | NM_003712.3 | NP_003703.1 | ||
| NM_177526.2 | NP_803545.1 | |||
| NM_177543.2 | NP_808211.1 | |||
| XM_011528396.2 | XP_011526698.1 | |||
| AF035959.1 | AAC15968.1 | |||
| AF047760.1 | AAC32104.1 | |||
| AF056083.1 | AAC25666.1 | |||
| BC002806.2 | AAH02806.1 | |||
| BE615792.1 | ||||
| BG831889.1 | ||||
| BM050291.1 | ||||
| BT007021.1 | AAP35667.1 | |||
| HY160303.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833702 | Chr.19:238657 - 402129 on Build GRCh38 | Loss |
|
| nsv953937 | Chr.19:276901 - 301700 on Build GRCh38 | Deletion |
|
| nsv428360 | Chr.19:232069 - 702131 on Build GRCh38 | Gain |
|
| dgv3419n100 | Chr.19:90898 - 305459 on Build GRCh38 | Gain |
|
| esv3643342 | Chr.19:266428 - 341459 on Build GRCh38 | Gain |
|
| esv3643344 | Chr.19:286841 - 307454 on Build GRCh38 | Gain |
|
| nsv1058440 | Chr.19:255818 - 301668 on Build GRCh38 | Gain |
|
| esv2422333 | Chr.19:251718 - 404517 on Build GRCh38 | Duplication |
|
| esv2763195 | Chr.19:90910 - 296458 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114778907] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map