Assay Details
Target Gene Details
Entrez Gene ID: | 390916 |
Gene Name: | nudix hydrolase 19 |
Gene Aliases: |
RP2 |
Location: |
Chr.19:32691607-32713796 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT19 | NM_001105570.1 | 1 | 315 | NP_001099040.1 |
| XM_017026806.1 | 1 | 669 | XP_016882295.1 | |
| DN998957.1 | 1 | 304 |
More Information
Set Membership: |
|

Genomic Map