Assay Details
Target Gene Details
Entrez Gene ID: | 11133 |
Gene Name: | kaptin, actin binding protein |
Gene Aliases: |
2E4, MRT41 |
Location: |
Chr.19:47475141-47486792 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KPTN | NM_001291296.1 | NP_001278225.1 | ||
| NM_007059.3 | NP_008990.2 | |||
| NR_111923.1 | ||||
| XM_011526399.2 | XP_011524701.1 | |||
| XM_017026226.1 | XP_016881715.1 | |||
| XM_017026227.1 | XP_016881716.1 | |||
| XM_017026228.1 | XP_016881717.1 | |||
| XM_017026229.1 | XP_016881718.1 | |||
| AF105369.1 | AAD39358.1 | |||
| AK023996.1 | ||||
| AK225419.1 | ||||
| AK298672.1 | ||||
| BC009249.2 | AAH09249.1 | |||
| DA129265.1 | ||||
| DA824599.1 | ||||
| DR759729.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758765 | Chr.19:47354499 - 47522248 on Build GRCh38 | Loss |
|
| nsv1056887 | Chr.19:47481963 - 47527369 on Build GRCh38 | Gain |
|
| nsv579888 | Chr.19:47428355 - 47658874 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map