Assay Details
Target Gene Details
Entrez Gene ID: | 1785 |
Gene Name: | dynamin 2 |
Gene Aliases: |
CMT2M, CMTDI1, CMTDIB, DI-CMTB, DYN2, DYNII, LCCS5 |
Location: |
Chr.19:10718053-10831910 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNM2 | NM_001005360.2 | NP_001005360.1 | ||
| NM_001005361.2 | NP_001005361.1 | |||
| NM_001005362.2 | NP_001005362.1 | |||
| NM_001190716.1 | NP_001177645.1 | |||
| NM_004945.3 | NP_004936.2 | |||
| AB209213.1 | BAD92450.1 | |||
| AK289831.1 | ||||
| AK312260.1 | ||||
| BC039596.1 | AAH39596.1 | |||
| BC054501.1 | AAH54501.1 | |||
| BM148999.1 | ||||
| DB069057.1 | ||||
| L36983.1 | AAA88025.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160584 | Chr.19:10618877 - 10892186 on Build GRCh38 | Deletion |
|
| nsv833746 | Chr.19:10678751 - 10842756 on Build GRCh38 | Loss |
|
| nsv1160585 | Chr.19:10674243 - 10771056 on Build GRCh38 | Deletion |
|
| nsv1061882 | Chr.19:10704200 - 10737778 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map