Assay Details
Target Gene Details
Entrez Gene ID: | 388553 |
Gene Name: | basic helix-loop-helix and HMG-box containing 1 |
Gene Aliases: |
HMGDC |
Location: |
Chr.19:45733251-45764692 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BHMG1 | NM_001310124.1 | NP_001297053.1 | ||
| XM_011527571.2 | XP_011525873.1 | |||
| XM_011527573.2 | XP_011525875.1 | |||
| XM_011527574.2 | XP_011525876.1 | |||
| DV080302.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv579870 | Chr.19:45704552 - 45814335 on Build GRCh38 | Loss |
|
| esv987735 | Chr.19:45742165 - 45761112 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map