Assay Details
Target Gene Details
Entrez Gene ID: | 100529264 |
Gene Name: | RAB4B-EGLN2 readthrough (NMD candidate) |
Gene Aliases: |
RERT-lncRNA |
Location: |
Chr.19:40778219-40808441 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB4B-EGLN2 | NR_037791.1 | |||
| AK291385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833832 | Chr.19:40754458 - 40828692 on Build GRCh38 | Loss |
|
| esv2758762 | Chr.19:40785722 - 41040163 on Build GRCh38 | Gain+Loss |
|

Genomic Map