Assay Details
Target Gene Details
Entrez Gene ID: | 59284 |
Gene Name: | calcium voltage-gated channel auxiliary subunit gamma 7 |
Gene Aliases: |
- |
Location: |
Chr.19:53909425-53943944 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNG7 | XM_005259124.3 | XP_005259181.1 | ||
| XM_017027093.1 | XP_016882582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064558 | Chr.19:53773468 - 53957491 on Build GRCh38 | Gain |
|
| nsv2541 | Chr.19:53901764 - 53946589 on Build GRCh38 | Deletion |
|
| esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion |
|
| nsv833876 | Chr.19:53845956 - 53995117 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map