Assay Details
Target Gene Details
Entrez Gene ID: | 8775 |
Gene Name: | NSF attachment protein alpha |
Gene Aliases: |
SNAPA |
Location: |
Chr.19:47487634-47515258 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAPA | NM_003827.3 | NP_003818.2 | ||
| NR_038456.1 | ||||
| NR_038457.1 | ||||
| XM_011527436.1 | XP_011525738.1 | |||
| XM_011527437.1 | XP_011525739.1 | |||
| AK098401.1 | ||||
| AK292244.1 | ||||
| AK308633.1 | ||||
| AK310114.1 | ||||
| AK310185.1 | ||||
| AY927608.1 | ||||
| AY927609.1 | ||||
| BC001165.1 | AAH01165.1 | |||
| BC007432.2 | AAH07432.1 | |||
| BC028234.1 | AAH28234.1 | |||
| BC091511.1 | AAH91511.1 | |||
| BQ948081.1 | ||||
| BT019568.1 | AAV38375.1 | |||
| CX873074.1 | ||||
| U39412.1 | AAC80170.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758765 | Chr.19:47354499 - 47522248 on Build GRCh38 | Loss |
|
| nsv1056887 | Chr.19:47481963 - 47527369 on Build GRCh38 | Gain |
|
| nsv579888 | Chr.19:47428355 - 47658874 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114700131] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map