Assay Details
Target Gene Details
Entrez Gene ID: | 125950 |
Gene Name: | ribonucleoprotein, PTB binding 1 |
Gene Aliases: |
- |
Location: |
Chr.19:10316212-10333638 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAVER1 | NM_133452.2 | NP_597709.2 | ||
| XM_011527671.1 | XP_011525973.1 | |||
| AB075858.1 | BAB85564.1 | |||
| BC037565.1 | AAH37565.1 | |||
| BX401103.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470124 | Chr.19:10289228 - 10362257 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map