Assay Details
Target Gene Details
Entrez Gene ID: | 390916 |
Gene Name: | nudix hydrolase 19 |
Gene Aliases: |
RP2 |
Location: |
Chr.19:32691607-32713796 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT19 | NM_001105570.1 | NP_001099040.1 | ||
| DA658958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv579311 | Chr.19:32708877 - 32720911 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140331296] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map