Assay Details
Target Gene Details
Entrez Gene ID: | 148170 |
Gene Name: | CDC42 effector protein 5 |
Gene Aliases: |
Borg3, CEP5 |
Location: |
Chr.19:54465025-54473290 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC42EP5 | NM_145057.3 | NP_659494.2 | ||
| BC024327.1 | AAH24327.1 | |||
| BC065910.1 | AAH65910.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3663n100 | Chr.19:54289334 - 54592227 on Build GRCh38 | Gain |
|
| esv2758771 | Chr.19:54163847 - 54954956 on Build GRCh38 | Gain+Loss |
|
| nsv833878 | Chr.19:54268967 - 54518890 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map