Assay Details
Target Gene Details
Entrez Gene ID: | 58509 |
Gene Name: | cactin, spliceosome C complex subunit |
Gene Aliases: |
C19orf29, NY-REN-24, fSAPc |
Location: |
Chr.19:3610628-3626815 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACTIN | NM_001080543.1 | 9 | 1672 | NP_001074012.1 |
| NM_021231.1 | 9 | 1672 | NP_067054.1 | |
| AF155102.1 | AAD42868.1 | |||
| AY917150.1 | 9 | 1674 | ||
| BC019848.1 | 2 | 216 | AAH19848.1 |
Target Gene Details
Entrez Gene ID: | 404665 |
Gene Name: | CACTIN antisense RNA 1 |
Gene Aliases: |
C19orf29-AS1, C19orf29OS |
Location: |
Chr.19:3607247-3613930 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACTIN-AS1 | NR_038865.1 | 4 | 1208 | |
| AK097999.1 | 4 | 1209 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953948 | Chr.19:3171703 - 3700002 on Build GRCh38 | Deletion |
|
| nsv833713 | Chr.19:3564423 - 3627361 on Build GRCh38 | Loss |
|
| nsv470114 | Chr.19:3372516 - 3855979 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map