Assay Details
Target Gene Details
Entrez Gene ID: | 342977 |
Gene Name: | nanos C2HC-type zinc finger 3 |
Gene Aliases: |
NANOS1L, NOS3, ZC2HC12C |
Location: |
Chr.19:13876383-13880762 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NANOS3 | NM_001098622.2 | 1 | 300 | NP_001092092.1 |
| XM_011527964.2 | 1 | 1166 | XP_011526266.1 | |
| BC101209.2 | 1 | 303 | AAI01210.1 | |
| BC101210.1 | 1 | 303 | ||
| DN993610.1 | 1 | 421 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833760 | Chr.19:13872472 - 13928073 on Build GRCh38 | Loss |
|
| nsv953979 | Chr.19:13795787 - 14006888 on Build GRCh38 | Deletion |
|
| nsv833757 | Chr.19:13725904 - 13951988 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map