Assay Details
Target Gene Details
Entrez Gene ID: | 284352 |
Gene Name: | protein phosphatase 1 regulatory subunit 37 |
Gene Aliases: |
LRRC68 |
Location: |
Chr.19:45093173-45147286 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R37 | NM_019121.1 | NP_061994.1 | ||
| XM_006723159.2 | XP_006723222.1 | |||
| BC035704.1 | AAH35704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953587 | Chr.19:45039143 - 45166142 on Build GRCh38 | Deletion |
|
| nsv833843 | Chr.19:45090630 - 45290424 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map