Assay Details
Target Gene Details
Entrez Gene ID: | 2906 |
Gene Name: | glutamate ionotropic receptor NMDA type subunit 2D |
Gene Aliases: |
EB11, GluN2D, NMDAR2D, NR2D |
Location: |
Chr.19:48394875-48444937 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIN2D | NM_000836.2 | NP_000827.2 | ||
| XM_011526872.1 | XP_011525174.1 | |||
| U77783.1 | AAC15910.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv833854 | Chr.19:48250143 - 48424167 on Build GRCh38 | Loss |
|
| esv34181 | Chr.19:48009377 - 48416682 on Build GRCh38 | Loss |
|
| nsv828572 | Chr.19:48399035 - 48497609 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map