Assay Details
Target Gene Details
Entrez Gene ID: | 56917 |
Gene Name: | Meis homeobox 3 |
Gene Aliases: |
MRG2 |
Location: |
Chr.19:47403118-47422315 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEIS3 | XM_011527139.2 | XP_011525441.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057003 | Chr.19:47416492 - 47471403 on Build GRCh38 | Loss |
|
| esv2758765 | Chr.19:47354499 - 47522248 on Build GRCh38 | Loss |
|
| nsv517901 | Chr.19:47373953 - 47444971 on Build GRCh38 | Loss |
|
| nsv9738 | Chr.19:47356269 - 47427931 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map