Assay Details
Target Gene Details
Entrez Gene ID: | 4051 |
Gene Name: | cytochrome P450 family 4 subfamily F member 3 |
Gene Aliases: |
CPF3, CYP4F, LTB4H |
Location: |
Chr.19:15640897-15660760 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP4F3 | NM_000896.2 | NP_000887.2 | ||
| NM_001199208.1 | NP_001186137.1 | |||
| NM_001199209.1 | NP_001186138.1 | |||
| XM_005259911.4 | XP_005259968.1 | |||
| XM_011528014.2 | XP_011526316.1 | |||
| XM_017026814.1 | XP_016882303.1 | |||
| XM_017026815.1 | XP_016882304.1 | |||
| AB002454.1 | BAA25990.1 | |||
| AF054821.1 | AAC08589.1 | |||
| AK296761.1 | ||||
| AK296802.1 | ||||
| AK298675.1 | ||||
| AK303477.1 | ||||
| AK304200.1 | ||||
| AK316136.1 | ||||
| BC136299.1 | ||||
| D12620.1 | BAA02144.1 | |||
| D12621.1 | BAA02145.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470128 | Chr.19:15653261 - 15728555 on Build GRCh38 | Gain |
|
| esv3643775 | Chr.19:15646813 - 15666335 on Build GRCh38 | Loss |
|
| esv2718235 | Chr.19:15616082 - 15677993 on Build GRCh38 | Deletion |
|
| dgv627e199 | Chr.19:15615949 - 15676977 on Build GRCh38 | Deletion |
|
| nsv9670 | Chr.19:15649504 - 15665950 on Build GRCh38 | Loss |
|
| nsv578718 | Chr.19:15654151 - 15656056 on Build GRCh38 | Gain |
|
| nsv1064684 | Chr.19:15311319 - 16062011 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map