Assay Details
Target Gene Details
Entrez Gene ID: | 55423 |
Gene Name: | signal regulatory protein gamma |
Gene Aliases: |
CD172g, SIRP-B2, SIRPB2, SIRPgamma, bA77C3.1 |
Location: |
Chr.20:1629152-1686516 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRPG | NM_001039508.1 | NP_001034597.1 | ||
| NM_018556.3 | NP_061026.2 | |||
| NM_080816.2 | NP_543006.2 | |||
| XM_005260749.3 | XP_005260806.1 | |||
| XM_011529286.2 | XP_011527588.1 | |||
| XM_011529287.2 | XP_011527589.1 | |||
| AB042624.1 | BAA95692.1 | |||
| AK292745.1 | ||||
| AY748247.1 | AAV88530.1 | |||
| AY748248.1 | AAV88531.1 | |||
| BC020629.1 | ||||
| BC064532.1 | AAH64532.1 | |||
| DA008008.1 |
Target Gene Details
Entrez Gene ID: | 101929010 |
Gene Name: | SIRPG antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:1633508-1648473 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRPG-AS1 | NR_110090.1 | 3 | 1374 | |
| AK093519.1 | 3 | 1387 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522008 | Chr.20:1359746 - 1657932 on Build GRCh38 | Gain |
|
| esv2758777 | Chr.20:1496135 - 1698610 on Build GRCh38 | Gain+Loss |
|
| esv3644988 | Chr.20:1626418 - 1861080 on Build GRCh38 | Gain |
|
| nsv585206 | Chr.20:1405829 - 1733716 on Build GRCh38 | Gain |
|
| esv2722071 | Chr.20:1565136 - 1694217 on Build GRCh38 | Deletion |
|
| nsv833894 | Chr.20:1538787 - 1772720 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map