Assay Details
Target Gene Details
Entrez Gene ID: | 1457 |
Gene Name: | casein kinase 2 alpha 1 |
Gene Aliases: |
CK2A1, CKII, CSNK2A3 |
Location: |
Chr.20:482694-543838 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSNK2A1 | NM_001895.3 | NP_001886.1 | ||
| NM_177559.2 | NP_808227.1 | |||
| NM_177560.2 | NP_808228.1 | |||
| AB451279.1 | ||||
| AB451411.1 | ||||
| AK302583.1 | ||||
| AK312535.1 | ||||
| BC011668.2 | AAH11668.1 | |||
| BC050036.1 | AAH50036.1 | |||
| BC053532.1 | AAH53532.1 | |||
| BC071167.1 | AAH71167.1 | |||
| BT019792.1 | AAV38595.1 | |||
| DQ354583.1 | ABC86794.1 | |||
| J02853.1 | AAA56821.1 | |||
| M55265.1 | AAA35503.1 | |||
| S53149.1 | ABB72474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4221n100 | Chr.20:449912 - 546851 on Build GRCh38 | Gain |
|
| nsv1066598 | Chr.20:422749 - 503206 on Build GRCh38 | Gain |
|
| nsv469833 | Chr.20:447851 - 614798 on Build GRCh38 | Gain |
|
| esv3644953 | Chr.20:397290 - 502310 on Build GRCh38 | Gain |
|
| dgv4222n100 | Chr.20:483143 - 524933 on Build GRCh38 | Gain |
|
| esv3644952 | Chr.20:386013 - 1119183 on Build GRCh38 | Gain |
|
| nsv1056266 | Chr.20:401152 - 522755 on Build GRCh38 | Gain |
|
| nsv833891 | Chr.20:468248 - 620062 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map