Assay Details
Target Gene Details
Entrez Gene ID: | 140876 |
Gene Name: | family with sequence similarity 65 member C |
Gene Aliases: |
C20orf175, C20orf176 |
Location: |
Chr.20:50586108-50692004 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM65C | NM_001290268.1 | NP_001277197.1 | ||
| NR_110890.1 | ||||
| XM_005260294.3 | XP_005260351.1 | |||
| XM_006723713.3 | XP_006723776.1 | |||
| XM_011528578.2 | XP_011526880.1 | |||
| XM_011528579.2 | XP_011526881.1 | |||
| XM_011528580.2 | XP_011526882.1 | |||
| XM_011528581.2 | XP_011526883.1 | |||
| XM_011528584.2 | XP_011526886.1 | |||
| XM_011528586.2 | XP_011526888.1 | |||
| XM_017027682.1 | XP_016883171.1 | |||
| AK295781.1 | ||||
| AK299337.1 | ||||
| BM846795.1 | ||||
| DC400584.1 |
Target Gene Details
Entrez Gene ID: | 100506175 |
Gene Name: | uncharacterized LOC100506175 |
Gene Aliases: |
- |
Location: |
Chr.20:50645471-50660291 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506175 | NR_111906.1 | |||
| AA044156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834001 | Chr.20:50625617 - 50820418 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map