Assay Details
Target Gene Details
Entrez Gene ID: | 644524 |
Gene Name: | NK2 homeobox 4 |
Gene Aliases: |
NKX2.4, NKX2D |
Location: |
Chr.20:21395367-21397409 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX2-4 | NM_033176.1 | 1 | 40 | NP_149416.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828702 | Chr.20:21395717 - 21397449 on Build GRCh38 | Gain |
|
| nsv1143980 | Chr.20:21396062 - 21397762 on Build GRCh38 | Deletion |
|
| nsv833941 | Chr.20:21393666 - 21576223 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map