Assay Details
Target Gene Details
Entrez Gene ID: | 128859 |
Gene Name: | BPI fold containing family B member 6 |
Gene Aliases: |
BPIL3, LPLUNC6 |
Location: |
Chr.20:33031648-33044108 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPIFB6 | NM_174897.2 | NP_777557.1 | ||
| XM_017027663.1 | XP_016883152.1 | |||
| AF465767.1 | AAM73985.1 | |||
| AY927631.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3567952 | Chr.20:33028987 - 33034277 on Build GRCh38 | Loss |
|
| nsv1056411 | Chr.20:32469234 - 33355029 on Build GRCh38 | Loss |
|
| esv2758792 | Chr.20:32908001 - 33160360 on Build GRCh38 | Loss |
|
| dgv4301n100 | Chr.20:33026982 - 33050770 on Build GRCh38 | Gain |
|
| nsv3351 | Chr.20:33014295 - 33037533 on Build GRCh38 | Insertion |
|
| nsv3352 | Chr.20:33022133 - 33066950 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map