Assay Details
Target Gene Details
Entrez Gene ID: | 671 |
Gene Name: | bactericidal/permeability-increasing protein |
Gene Aliases: |
BPIFD1, rBPI |
Location: |
Chr.20:38304150-38337503 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPI | NM_001725.2 | NP_001716.2 | ||
| AF322588.1 | AAG42844.1 | |||
| AK296568.1 | ||||
| AK315328.1 | ||||
| AX713278.1 | CAD88641.1 | |||
| AX746123.1 | CAD97475.1 | |||
| AX752831.1 | CAD99178.1 | |||
| BC040955.1 | AAH40955.1 | |||
| BP306215.1 | ||||
| DQ414688.1 | ABD66755.1 | |||
| J04739.1 | AAA51841.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058457 | Chr.20:38277492 - 38844429 on Build GRCh38 | Loss |
|
| nsv585964 | Chr.20:38286018 - 38343306 on Build GRCh38 | Loss |
|
| esv35121 | Chr.20:38305228 - 38453502 on Build GRCh38 | Gain |
|
| nsv1063495 | Chr.20:38295280 - 38457885 on Build GRCh38 | Gain |
|
| dgv4309n100 | Chr.20:38271961 - 38345891 on Build GRCh38 | Loss |
|
| esv3645775 | Chr.20:38264344 - 38392152 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map